Assay Design



Gene Transcripts

Gene Symbol
Ip6k3
Entrez Gene ID
100772883
Gene Name
inositol hexakisphosphate kinase 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614972: 80445 - 102442 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510823.2 XP_003510871.1 2 - 3 463 62
XM_007651405.1 XP_007649595.1 2 - 3 384 62

View Full Product Details