Assay Design



Gene Transcripts

Gene Symbol
Zdhhc11b
Entrez Gene ID
100762396
Gene Name
zinc finger, DHHC-type containing 11B
Gene Aliases
I79_023049
Chromosome Location
Chr.Un|NW_003616237: 4368 - 21885 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513952.1 XP_003514000.1 2 - 3 323 56

View Full Product Details