Assay Design



Gene Transcripts

Gene Symbol
Adra2b
Entrez Gene ID
100757384
Gene Name
adrenoceptor alpha 2B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613665: 2008654 - 2010003 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007618099.1 XP_007616289.1 1 - 1 1220 62

View Full Product Details