Assay Design



Gene Transcripts

Gene Symbol
Rsph6a
Entrez Gene ID
100758087
Gene Name
radial spoke head 6 homolog A (Chlamydomonas)
Gene Aliases
I79_012668
Chromosome Location
Chr.Un|NW_003614150: 875876 - 904798 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645980.1 XP_007644170.1 5 - 6 815 82

View Full Product Details