Assay Design



Gene Transcripts

Gene Symbol
Rwdd4
Entrez Gene ID
100752671
Gene Name
RWD domain containing 4
Gene Aliases
I79_000711
Chromosome Location
Chr.Un|NW_003613593: 2207065 - 2226853 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495468.2 XP_003495516.1 2 - 3 183 100
XM_007641274.1 XP_007639464.1 2 - 3 183 100
XM_007641285.1 XP_007639475.1 2 - 3 183 100
GenBank mRNA JI892731.1 - 6 - 7 100

View Full Product Details