Assay Design



Gene Transcripts

Gene Symbol
Slc16a9
Entrez Gene ID
100760512
Gene Name
solute carrier family 16, member 9
Gene Aliases
I79_016926
Chromosome Location
Chr.Un|NW_003614603: 179704 - 205819 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508886.1 XP_003508934.1 2 - 3 341 104

View Full Product Details