Assay Design



Gene Transcripts

Gene Symbol
Bcdin3d
Entrez Gene ID
100757267
Gene Name
BCDIN3 domain containing
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616024: 74847 - 79665 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513597.2 XP_003513645.2 1 - 2 240 98

View Full Product Details