Assay Design



Gene Transcripts

Gene Symbol
Med30
Entrez Gene ID
100751133
Gene Name
mediator complex subunit 30
Gene Aliases
I79_003555
Chromosome Location
Chr.Un|NW_003613668: 103035 - 114604 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007619112.1 XP_007617302.1 1 - 1 339 71
GenBank mRNA JI882403.1 - 1 - 1 339 71

View Full Product Details