Assay Design



Gene Transcripts

Gene Symbol
Dact1
Entrez Gene ID
100768384
Gene Name
dishevelled-binding antagonist of beta-catenin 1
Gene Aliases
I79_022994
Chromosome Location
Chr.Un|NW_003616202: 51577 - 60458 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654127.1 XP_007652317.1 2 - 3 263 66

View Full Product Details