Assay Design



Gene Transcripts

Gene Symbol
Hoxa3
Entrez Gene ID
100761845
Gene Name
homeobox A3
Gene Aliases
I79_008448
Chromosome Location
Chr.Un|NW_003613870: 22344 - 64408 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642616.1 XP_007640806.1 5 - 6 1154 79

View Full Product Details