Assay Design



Gene Transcripts

Gene Symbol
Samd10
Entrez Gene ID
100764555
Gene Name
sterile alpha motif domain containing 10
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613943: 1548811 - 1552719 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503145.2 XP_003503193.1 3 - 4 518 75
XM_007643785.1 XP_007641975.1 3 - 4 518 75

View Full Product Details