Assay Design



Gene Transcripts

Gene Symbol
Ms4a13
Entrez Gene ID
100772510
Gene Name
membrane-spanning 4-domains, subfamily A, member 13
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617032: 10620 - 29371 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514844.2 XP_003514892.1 2 - 3 235 86

View Full Product Details