Assay Design



Gene Transcripts

Gene Symbol
Ccrn4l
Entrez Gene ID
100766002
Gene Name
CCR4 carbon catabolite repression 4-like (S. cerevisiae)
Gene Aliases
I79_007533, Nocturnin
Chromosome Location
Chr.Un|NW_003613834: 1098031 - 1119244 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641860.1 XP_007640050.1 2 - 3 303 72
GenBank mRNA JI896616.1 - 1 - 2 264 72

View Full Product Details