Assay Design



Gene Transcripts

Gene Symbol
Ensa
Entrez Gene ID
100764533
Gene Name
endosulfine alpha
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613699: 1775107 - 1790926 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498650.2 XP_003498698.1 3 - 4 492 53

View Full Product Details