Assay Design
Gene Transcripts
- Gene Symbol
- Prkrir
- Entrez Gene ID
-
100750790
- Gene Name
- protein-kinase, interferon-inducible double stranded RNA dependent inhibitor, repressor of (P58 repressor)
- Gene Aliases
-
I79_014684
- Chromosome Location
-
Chr.Un|NW_003614327: 374614 - 391033 on Build CriGri-PICRH-1.0
View Full Product Details