Assay Design



Gene Transcripts

Gene Symbol
Fgfbp1
Entrez Gene ID
100762110
Gene Name
fibroblast growth factor binding protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617915: 40027 - 42742 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515381.2 XP_003515429.1 2 - 2 585 69

View Full Product Details