Assay Design



Gene Transcripts

Gene Symbol
Rab11fip1
Entrez Gene ID
100750840
Gene Name
RAB11 family interacting protein 1 (class I)
Gene Aliases
I79_003990
Chromosome Location
Chr.Un|NW_003613685: 501960 - 522748 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498235.1 XP_003498283.1 4 - 5 3052 68

View Full Product Details