Assay Design



Gene Transcripts

Gene Symbol
Pacrg
Entrez Gene ID
100751258
Gene Name
PARK2 co-regulated
Gene Aliases
I79_006725
Chromosome Location
Chr.Un|NW_003613794: 346197 - 762449 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500467.2 XP_003500515.1 4 - 5 964 97

View Full Product Details