Assay Design



Gene Transcripts

Gene Symbol
Zbtb44
Entrez Gene ID
100754935
Gene Name
zinc finger and BTB domain containing 44
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613602: 4177532 - 4204215 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495841.2 XP_003495889.1 2 - 3 1159 95
XM_007644499.1 XP_007642689.1 2 - 3 1159 95
GenBank mRNA JI884593.1 - 2 - 3 505 95
JP054259.1 - 2 - 3 505 95

View Full Product Details