Assay Design



Gene Transcripts

Gene Symbol
Tmem95
Entrez Gene ID
100765490
Gene Name
transmembrane protein 95
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613643: 1932223 - 1934192 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497187.2 XP_003497235.1 5 - 6 589 77

View Full Product Details