Assay Design



Gene Transcripts

Gene Symbol
Spink14
Entrez Gene ID
100756891
Gene Name
serine peptidase inhibitor, Kazal type 14 (putative)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616611: 101318 - 105488 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514445.1 XP_003514493.1 2 - 3 109 117

View Full Product Details