Assay Design



Gene Transcripts

Gene Symbol
Thumpd1
Entrez Gene ID
100754766
Gene Name
THUMP domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617609: 4324 - 10724 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515248.2 XP_003515296.1 3 - 4 687 147
GenBank mRNA JI864165.1 - 3 - 4 513 147
JP050655.1 - 3 - 4 512 147

View Full Product Details