Assay Design



Gene Transcripts

Gene Symbol
Mblac2
Entrez Gene ID
100767767
Gene Name
metallo-beta-lactamase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614058: 435100 - 461257 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504393.2 XP_003504441.1 1 - 2 572 78

View Full Product Details