Assay Design



Gene Transcripts

Gene Symbol
Surf6
Entrez Gene ID
100771435
Gene Name
surfeit 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615220: 146002 - 151773 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511729.2 XP_003511777.1 3 - 4 465 96
GenBank mRNA JI883990.1 - 3 - 4 452 96
JP055378.1 - 3 - 4 452 96

View Full Product Details