Assay Design



Gene Transcripts

Gene Symbol
Adora1
Entrez Gene ID
100751346
Gene Name
adenosine A1 receptor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613706: 81915 - 114027 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498843.2 XP_003498891.1 1 - 2 340 62

View Full Product Details