Assay Design



Gene Transcripts

Gene Symbol
Tlr6
Entrez Gene ID
100689185
Gene Name
toll-like receptor 6
Gene Aliases
I79_025539
Chromosome Location
Chr.Un|NW_003619946: 11430 - 16743 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515801.2 XP_003515849.2 2 - 3 347 81
GenBank mRNA AF548356.1 - 1 - 2 128 81

View Full Product Details