Assay Design



Gene Transcripts

Gene Symbol
Nkx2-5
Entrez Gene ID
100774866
Gene Name
NK2 homeobox 5
Gene Aliases
I79_007135
Chromosome Location
Chr.Un|NW_003613815: 1291108 - 1297201 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641566.1 XP_007639756.1 4 - 5 692 86

View Full Product Details