Assay Design



Gene Transcripts

Gene Symbol
Lmod3
Entrez Gene ID
100755909
Gene Name
leiomodin 3 (fetal)
Gene Aliases
I79_000372
Chromosome Location
Chr.Un|NW_003613585: 4912476 - 4927283 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495210.1 XP_003495258.1 1 - 2 297 73

View Full Product Details