Assay Design



Gene Transcripts

Gene Symbol
Mb21d2
Entrez Gene ID
100750802
Gene Name
Mab-21 domain containing 2
Gene Aliases
I79_018532
Chromosome Location
Chr.Un|NW_003614854: 11764 - 22094 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510267.1 XP_003510315.1 3 - 4 237 80

View Full Product Details