Assay Design



Gene Transcripts

Gene Symbol
Lrrtm4
Entrez Gene ID
100767641
Gene Name
leucine rich repeat transmembrane neuronal 4
Gene Aliases
I79_003717
Chromosome Location
Chr.Un|NW_003613675: 1713513 - 2502450 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007620956.1 XP_007619146.1 1 - 2 110 95

View Full Product Details