Assay Design



Gene Transcripts

Gene Symbol
Crhbp
Entrez Gene ID
100757291
Gene Name
corticotropin releasing hormone binding protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613700: 1858389 - 1886380 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498707.2 XP_003498755.1 7 - 8 1393 76
XM_007632075.1 XP_007630265.1 8 - 9 1272 76

View Full Product Details