Assay Design



Gene Transcripts

Gene Symbol
Bend4
Entrez Gene ID
100766260
Gene Name
BEN domain containing 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616124: 4112 - 24982 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513778.2 XP_003513826.1 1 - 2 606 86
GenBank mRNA JI886972.1 - 1 - 2 542 86
JP059225.1 - 1 - 2 542 86

View Full Product Details