Assay Design



Gene Transcripts

Gene Symbol
Nrip2
Entrez Gene ID
100769051
Gene Name
nuclear receptor interacting protein 2
Gene Aliases
I79_001347
Chromosome Location
Chr.Un|NW_003613610: 313850 - 322804 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496017.2 XP_003496065.1 1 - 2 330 73
XM_007646261.1 XP_007644451.1 1 - 2 330 73

View Full Product Details