Assay Design



Gene Transcripts

Gene Symbol
Ankrd33b
Entrez Gene ID
100760181
Gene Name
ankyrin repeat domain 33B
Gene Aliases
I79_006343
Chromosome Location
Chr.Un|NW_003613775: 1631197 - 1694590 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500138.1 XP_003500186.1 5 - 6 542 67

View Full Product Details