Assay Design



Gene Transcripts

Gene Symbol
Reep5
Entrez Gene ID
100753505
Gene Name
receptor accessory protein 5
Gene Aliases
I79_009156
Chromosome Location
Chr.Un|NW_003613902: 1031883 - 1042734 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643150.1 XP_007641340.1 2 - 3 551 75
GenBank mRNA JI863564.1 - 2 - 3 551 75

View Full Product Details