Assay Design



Gene Transcripts

Gene Symbol
Ybey
Entrez Gene ID
100752720
Gene Name
ybeY metallopeptidase (putative)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613799: 289573 - 295811 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500548.2 XP_003500596.1 3 - 4 375 80
XM_007641319.1 XP_007639509.1 3 - 4 595 80

View Full Product Details