Assay Design



Gene Transcripts

Gene Symbol
Lrrc10b
Entrez Gene ID
100771537
Gene Name
leucine rich repeat containing 10B
Gene Aliases
I79_022211
Chromosome Location
Chr.Un|NW_003615831: 95211 - 95629 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653577.1 XP_007651767.1 1 - 1 317 64

View Full Product Details