Assay Design



Gene Transcripts

Gene Symbol
Vmo1
Entrez Gene ID
100769165
Gene Name
vitelline membrane outer layer 1 homolog (chicken)
Gene Aliases
I79_002790
Chromosome Location
Chr.Un|NW_003613643: 2539806 - 2540890 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497199.2 XP_003497247.1 2 - 3 311 81

View Full Product Details