Assay Design



Gene Transcripts

Gene Symbol
Aadacl3
Entrez Gene ID
100751803
Gene Name
arylacetamide deacetylase-like 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616544: 84230 - 91612 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514360.1 XP_003514408.1 3 - 4 448 64

View Full Product Details