Assay Design



Gene Transcripts

Gene Symbol
Mlana
Entrez Gene ID
100762259
Gene Name
melan-A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614848: 438596 - 447202 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510223.1 XP_003510271.1 2 - 3 164 94

View Full Product Details