Assay Design



Gene Transcripts

Gene Symbol
Zfp91
Entrez Gene ID
100751041
Gene Name
ZFP91 zinc finger protein
Gene Aliases
I79_023719
Chromosome Location
Chr.Un|NW_003616654: 65268 - 97840 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654606.1 XP_007652796.1 4 - 5 362 103
GenBank mRNA JP055359.1 - 4 - 5 354 103

View Full Product Details