Assay Design



Gene Transcripts

Gene Symbol
Edaradd
Entrez Gene ID
100765451
Gene Name
EDAR-associated death domain
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614408: 529646 - 572746 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648180.1 XP_007646370.1 3 - 4 603 83

View Full Product Details