Assay Design



Gene Transcripts

Gene Symbol
Nme3
Entrez Gene ID
100752009
Gene Name
NME/NM23 nucleoside diphosphate kinase 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613852: 1396512 - 1397568 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501491.2 XP_003501539.1 3 - 4 313 93
XM_007642188.1 XP_007640378.1 3 - 4 298 93
GenBank mRNA JI896363.1 - 2 - 3 125 93
JP055150.1 - 2 - 3 125 93

View Full Product Details