Assay Design



Gene Transcripts

Gene Symbol
Hpcal1
Entrez Gene ID
100752247
Gene Name
hippocalcin-like 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614717: 86507 - 110131 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509475.2 XP_003509523.1 2 - 3 493 60
GenBank mRNA JI885858.1 - 1 - 2 155 60
JP052220.1 - 1 - 2 154 60

View Full Product Details