Assay Design



Gene Transcripts

Gene Symbol
Phlda3
Entrez Gene ID
100764336
Gene Name
pleckstrin homology-like domain, family A, member 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613706: 1598814 - 1601368 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498886.2 XP_003498934.1 1 - 2 315 97

View Full Product Details