Assay Design



Gene Transcripts

Gene Symbol
Ceacam19
Entrez Gene ID
100750495
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 19
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614150: 63130 - 90568 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505458.2 XP_003505506.2 2 - 3 424 69

View Full Product Details