Assay Design



Gene Transcripts

Gene Symbol
Vma21
Entrez Gene ID
100772090
Gene Name
VMA21 vacuolar H+-ATPase homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614816: 192552 - 201659 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510037.2 XP_003510085.1 2 - 3 275 113
GenBank mRNA JI886243.1 - 2 - 3 268 113
JP053937.1 - 2 - 3 268 113

View Full Product Details