Assay Design



Gene Transcripts

Gene Symbol
Cmc2
Entrez Gene ID
100764782
Gene Name
C-x(9)-C motif containing 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614582: 500515 - 528980 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508770.2 XP_003508818.1 2 - 3 201 90
XM_007649394.1 XP_007647584.1 2 - 3 164 90

View Full Product Details