Assay Design



Gene Transcripts

Gene Symbol
Catsperb
Entrez Gene ID
100751204
Gene Name
catsper channel auxiliary subunit beta
Gene Aliases
I79_020434
Chromosome Location
Chr.Un|NW_003615240: 63924 - 189665 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511823.1 XP_003511871.1 6 - 7 615 91

View Full Product Details