Assay Design



Gene Transcripts

Gene Symbol
Espnl
Entrez Gene ID
100773927
Gene Name
espin-like
Gene Aliases
I79_017467
Chromosome Location
Chr.Un|NW_003614689: 365850 - 389372 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649917.1 XP_007648107.1 7 - 8 1215 95

View Full Product Details